MAT491652590 Matrix Scientific

5-(Dimethylamino)naphthalene-1-sulfonic acid

  • MDL number MFCD00003995
  • Cas Number : 4272-77-9
  • Molecular Weight : 251.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/146013.html

Safety Information

MSDS http://www.matrixscientific.com/146013.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart