MAT491656914 Matrix Scientific

2-Chloro-3-fluoropyridine 1-oxide

  • MDL number MFCD06617948
  • Cas Number : 85386-94-3
  • Molecular Weight : 147.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/150234.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart