MAT491656946 Matrix Scientific

2-Chloro-6-methyl-4-nitropyridine

  • MDL number MFCD14582081
  • Cas Number : 79055-51-9
  • Molecular Weight : 172.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/150266.html

Safety Information

MSDS http://www.matrixscientific.com/150266.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart