MAT491657389 Matrix Scientific

2,4-Dibromopyridin-3-ol

  • MDL number MFCD11044315
  • Cas Number : 129611-31-0
  • Molecular Weight : 252.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/150688.html

Safety Information

MSDS http://www.matrixscientific.com/150688.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart