MAT491659760 Matrix Scientific

Ethyl 4-amino-3-oxobutanoate hydrochloride

  • MDL number MFCD20693743
  • Cas Number : 86578-58-7
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/153046.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/153046.html
Availability details for
Quantity
  • Availability
  • Add to Cart