MAT491660062 Matrix Scientific

2-Benzyl-2,7-diazaspiro[3.5]nonane

  • MDL number MFCD11227023
  • Cas Number : 2177265-01-7
  • Molecular Weight : 252.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/153348.html

Safety Information

MSDS http://www.matrixscientific.com/153348.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart