MAT502727084 Matrix Scientific

Ethyl 2-chloroethanimidoate hydrochloride

  • MDL number MFCD10697958
  • Cas Number : 36743-66-5
  • Molecular Weight : 158.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient/argon
Country of Origin USA
Product Link http://www.matrixscientific.com/154829.html

Safety Information

hcodes Irritant/Moisture Sensitive
Availability details for
Quantity
  • Availability
  • Add to Cart