MAT502727095 Matrix Scientific

1-[1-(2-Fluorophenyl)-1H-pyrazol-4-yl]ethanamine hydrochloride

  • MDL number MFCD09431461
  • Cas Number : 1049791-50-5
  • Molecular Weight : 241.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030926.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030926.html
Availability details for
Quantity
  • Availability
  • Add to Cart