MAT502727535 Matrix Scientific

Glycyltryptophan hydrate

  • MDL number MFCD27756519
  • Cas Number : 2390-74-1
  • Molecular Weight : 279.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/155280.html

Safety Information

MSDS http://www.matrixscientific.com/155280.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart