MAT502728126 Matrix Scientific

3-Isopropyl-1,1-dimethylindan-5-ol

  • MDL number MFCD02039507
  • Cas Number : 345992-03-2
  • Molecular Weight : 204.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/155867.html

Safety Information

MSDS http://www.matrixscientific.com/155867.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart