MAT502729528 Matrix Scientific

Dimethyl 2,2'-oxydiacetate

  • MDL number MFCD01026217
  • Cas Number : 7040-23-5
  • Molecular Weight : 162.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/157267.html

Safety Information

MSDS http://www.matrixscientific.com/157267.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart