MAT502731475 Matrix Scientific

N-{5-Chloro-4-[(4-chlorophenyl)(cyano)methyl]-2-methylphenyl}-2-hydroxybenzamide

  • MDL number MFCD27940093
  • Cas Number : 61439-05-2
  • Molecular Weight : 411.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/159276.html

Safety Information

MSDS http://www.matrixscientific.com/159276.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart