MAT502731541 Matrix Scientific

N-(Phenylsulfonyl)-L-tryptophan

  • MDL number MFCD09749744
  • Cas Number : 149586-34-5
  • Molecular Weight : 344.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/159342.html

Safety Information

MSDS http://www.matrixscientific.com/159342.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart