MAT502732649 Matrix Scientific

2-[(4-Isopropylphenyl)amino]nicotinic acid

  • MDL number MFCD03119359
  • Cas Number : 55285-34-2
  • Molecular Weight : 256.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/159651.html

Safety Information

MSDS http://www.matrixscientific.com/159651.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart