MAT502732666 Matrix Scientific

2-Isoxazol-5-ylpiperidine hydrochloride

  • MDL number MFCD29034986
  • Cas Number : 2098500-81-1
  • Molecular Weight : 188.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/160447.html

Safety Information

MSDS http://www.matrixscientific.com/160447.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart