MAT514455581 Matrix Scientific

3-(4-Fluorophenyl)-1,2,4-oxadiazole-5-carboxylic acid sodium salt

  • MDL number MFCD30181470
  • Cas Number : 944896-51-9
  • Molecular Weight : 230.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/099384.html

Safety Information

MSDS http://www.matrixscientific.com/099384.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart