MAT515993637 Matrix Scientific

N-[3-(Hydrazinocarbonyl)phenyl]-2-methylpropanamide

  • MDL number MFCD29277528
  • Cas Number : 2098500-83-3
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/160914.html

Safety Information

MSDS http://www.matrixscientific.com/160914.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart