MAT515993782 Matrix Scientific

2-[(2-Aminoethyl)thio]-N-(2,6-dimethylphenyl)-acetamide hydrochloride

  • MDL number MFCD00445670
  • Cas Number : 312926-99-1
  • Molecular Weight : 274.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037552.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037552.html
Availability details for
Quantity
  • Availability
  • Add to Cart