MAT517336494 Matrix Scientific

3-Bromo-5-iodopyridin-2-ol

  • MDL number MFCD11044276
  • Cas Number : 637348-81-3
  • Molecular Weight : 299.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/161803.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart