MAT842161771 Matrix Scientific

Ethyl 7-chloro-1-ethyl-6-nitro-4-oxo-1,4-dihydro-3-quinolinecarboxylate : ethanol (1:1)

  • MDL number MFCD29277529
  • Cas Number : 2098500-77-5
  • Molecular Weight : 370.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/059738.html

Safety Information

MSDS http://www.matrixscientific.com/059738.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart