MAT842181867 Matrix Scientific

Oxytocin

  • MDL number MFCD00076731
  • Cas Number : 50-56-6
  • Molecular Weight : 1007.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/168058.html

Safety Information

hcodes Irritant/Store in Freezer
Availability details for
Quantity
  • Availability
  • Add to Cart