MAT842189257 Matrix Scientific

3-(Dimethylamino)-1-(2-hydroxyphenyl)-2-propen-1-one

  • MDL number MFCD00172108
  • Cas Number : 106129-86-6
  • Molecular Weight : 191.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Product Link https://www.matrixscientific.com/169384.html

Safety Information

hcodes Irritant/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart