MAT842199241 Matrix Scientific

2-Bromo-4-methylpentane

  • MDL number MFCD00043446
  • Cas Number : 30310-22-6
  • Molecular Weight : 165.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 130-133
Product Link http://www.matrixscientific.com/172104.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/172104.html
Availability details for
Quantity
  • Availability
  • Add to Cart