MAT842203304 Matrix Scientific

Ethyl alpha-cyclopentylacetoacetate

  • MDL number MFCD00144276
  • Cas Number : 1540-32-5
  • Molecular Weight : 198.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Boiling point 125-130�mm
Product Link http://www.matrixscientific.com/172480.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart