MAT842203375 Matrix Scientific

5-Norbornene-2-methanol

  • MDL number MFCD00167571
  • Cas Number : 95-12-5
  • Molecular Weight : 124.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.03
Flash Point 87
Storage temp. Ambient
Country of Origin USA
Boiling point 120
Product Link http://www.matrixscientific.com/172551.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/172551.html
Availability details for
Quantity
  • Availability
  • Add to Cart