MATH03871FB6 Matrix Scientific

4-Carboxyl-L-phenylalanine hydrochloride

  • MDL number MFCD31381534
  • Cas Number : 126109-42-0
  • Molecular Weight : 245.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/089328.html

Safety Information

MSDS http://www.matrixscientific.com/089328.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart