MATH0A3B0613 Matrix Scientific

4-Chloro-2-fluorophenyl 4-piperidinylmethyl ether hydrochloride

  • MDL number MFCD31691561
  • Cas Number : 946680-87-1
  • Molecular Weight : 280.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023143.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023143.html
Availability details for
Quantity
  • Availability
  • Add to Cart