MATH0A3B06A0 Matrix Scientific

4-Methyl-4-phenylpentan-2-one

  • MDL number MFCD00089461
  • Cas Number : 7403-42-1
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/184526.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart