MATH3165D099 Matrix Scientific

Poly (hexamethylene biguanide), hydrochloride (as 20% aqueous solution)

  • MDL number MFCD00217750
  • Cas Number : 32289-58-0
  • Molecular Weight : 221.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/196142.html

Safety Information

MSDS http://www.matrixscientific.com/196142.html
Availability details for
Quantity
  • Availability
  • Add to Cart