MATH4642467F Matrix Scientific

17-Acetoxyestra-4,6-dien-3-one

  • MDL number MFCD00271529
  • Cas Number : 2590-41-2
  • Molecular Weight : 314.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/089229.html

Safety Information

MSDS http://www.matrixscientific.com/089229.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart