MATH46424681 Matrix Scientific

1,4-Dimethyl-1H-imidazol-2-amine hydrochloride

  • MDL number MFCD19208440
  • Cas Number : 2305252-87-1
  • Molecular Weight : 147.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/175983.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart