MATH8735F2BF Matrix Scientific

Endo-3-Amine-9-methyl-9-azabicyclo[3,3,1]nonane

  • MDL number MFCD07374273
  • Cas Number : 76272-56-5
  • Molecular Weight : 154.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/074391.html

Safety Information

MSDS http://www.matrixscientific.com/074391.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart