PRI424048072 Princeton BioMolecular Research, Inc.

2-(5-Chloro-2,7-dimethyl-1H-indol-3-yl)-ethylamine

  • MDL number MFCD02665973
  • Cas Number : 383146-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart