PRI424048102 Princeton BioMolecular Research, Inc.

5-Ethyl-3-methyl-benzofuran-2-carboxylic acid

  • MDL number MFCD06641569
  • Cas Number : 58455-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart