PRI424048171 Princeton BioMolecular Research, Inc.

1-(2-naphthyl)piperazine

  • MDL number MFCD03444412
  • Cas Number : 57536-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart