PRI424048191 Princeton BioMolecular Research, Inc.

6-methyl-1,2,4-triazine-3,5-diol

  • MDL number MFCD00127802
  • Molecular Weight : 127.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methyl-1,2,4-triazine-3,5-diol
Inchi Code 1S/C4H5N3O2/c1-2-3(8)5-4(9)7-6-2/h1H3,(H2,5,7,8,9)
Inchi Key XZWMZFQOHTWGQE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart