PRI424048214 Princeton BioMolecular Research, Inc.

4H-[1,2,4]Triazole-3-thiol

  • MDL number MFCD00456665
  • Cas Number : 3179-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart