PRI424048327 Princeton BioMolecular Research, Inc.

4-chloro-N-(3-methylphenyl)butanamide

  • MDL number MFCD00086969
  • Cas Number : 73863-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart