PRI424048367 Princeton BioMolecular Research, Inc.

5-(pyrrolidin-1-ylmethyl)-2-furoyl chloride

  • MDL number MFCD08436683
  • Cas Number : 914206-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart