PRI424048439 Princeton BioMolecular Research, Inc.

2-bromo-5-(4-ethoxyphenyl)-1,3,4-thiadiazole

  • MDL number MFCD08436701
  • Molecular Weight : 285.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(5-bromo-1,3,4-thiadiazol-2-yl)phenyl ethyl ether
Inchi Code 1S/C10H9BrN2OS/c1-2-14-8-5-3-7(4-6-8)9-12-13-10(11)15-9/h3-6H,2H2,1H3
Inchi Key TWSNJXHHIJIXCR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart