PRI424048448 Princeton BioMolecular Research, Inc.

1-ethyl-3,6-dimethyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD05667385
  • Cas Number : 832743-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart