PRI424048484 Princeton BioMolecular Research, Inc.

1-[(2,3-dichlorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04967888
  • Cas Number : 1004193-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart