PRI424048487 Princeton BioMolecular Research, Inc.

1-[(4-chloro-3-methylphenoxy)methyl]-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04967905
  • Cas Number : 957400-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart