PRI424048488 Princeton BioMolecular Research, Inc.

1-[(4-chloro-3,5-dimethylphenoxy)methyl]-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04967892
  • Cas Number : 1004193-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart