PRI424048492 Princeton BioMolecular Research, Inc.

1-[(4-chloro-3,5-dimethyl-1H-pyrazol-1-yl)methyl]-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04967758
  • Cas Number : 1004192-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart