PRI424048499 Princeton BioMolecular Research, Inc.

2-[3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

  • MDL number MFCD04969236
  • Cas Number : 1006328-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart