PRI424048519 Princeton BioMolecular Research, Inc.

5-[(4-methoxyphenoxy)methyl]-2-furoic acid

  • MDL number MFCD03421881
  • Cas Number : 832739-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart