PRI424048528 Princeton BioMolecular Research, Inc.

4-(4-chloro-3,5-dimethyl-1H-pyrazol-1-yl)aniline

  • MDL number MFCD08558192
  • Cas Number : 943750-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart