PRI424048536 Princeton BioMolecular Research, Inc.

1-(4-chloro-2-methylphenyl)hydrazine

  • MDL number MFCD02663074
  • Cas Number : 58791-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart