PRI424048591 Princeton BioMolecular Research, Inc.

1-(4-methylbenzyl)-1H-pyrazol-4-amine

  • MDL number MFCD02253995
  • Cas Number : 1002414-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart